missing translation for 'onlineSavingsMsg'
Learn More

CARD14, Rabbit, Polyclonal Antibody, Abnova™

Código de producto. 16057367
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16057367 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16057367 Proveedor Abnova N.º de proveedor PAB27180.100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit polyclonal antibody raised against synthetic peptide of CARD14.

The protein encoded by this gene belongs to the membrane-associated guanylate kinase (MAGUK) family, a class of proteins that functions as molecular scaffolds for the assembly of multiprotein complexes at specialized regions of the plasma membrane. This protein is also a member of the CARD protein family, which is defined by carrying a characteristic caspase-associated recruitment domain (CARD). This protein shares a similar domain structure with CARD11 protein. The CARD domains of both proteins have been shown to specifically interact with BCL10, a protein known to function as a positive regulator of cell apoptosis and NF-kappaB activation. When expressed in cells, this protein activated NF-kappaB and induced the phosphorylation of BCL10. Two alternatively spliced variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq

Especificaciones

Antígeno CARD14
Aplicaciones ELISA, Immunohistochemistry
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Descripción Rabbit polyclonal antibody raised against synthetic peptide of CARD14.
Dilución Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulación In PBS (0.02% sodium azide)
génica CARD14
N.º de referencia del gen NP_007715
Alias de gen BIMP2/CARMA2
Símbolos de los genes CARD14
Especie del huésped Rabbit
Inmunógeno A synthetic peptide corresponding to 14 amino acids at C-terminus of human CARD14.
Método de purificación Peptide affinity purification
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 79092
Especificidad de la prueba CARD14 antibody is human specific. At least three isoforms of CARD14 are known to exist; this antibody will only detect isoform 1. CARD14 antibody is predicted not to cross-react with other CARMA proteins.
Especies diana Human
Contenido y almacenamiento Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos

For Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.