Learn More
ANKK1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Descripción
The protein encoded by this gene belongs to the Ser/Thr protein kinase family, and protein kinase superfamily involved in signal transduction pathways. This gene is closely linked to DRD2 gene (GeneID:1813) on chr 11, and a well studied restriction fragment length polymorphism (RFLP) designated TaqIA, was originally associated with the DRD2 gene, however, later was determined to be located in exon 8 of ANKK1 gene (PMIDs: 18621654, 15146457), where it causes a nonconservative amino acid substitution. It is not clear if this gene plays any role in neuropsychiatric disorders previously associated with Taq1A RFLP. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | ANKK1 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Rabbit polyclonal antibody raised against synthetic peptide of ANKK1. |
| Dilución | Western Blot (1:500-1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user. |
| Formulación | In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) |
| génica | ANKK1 |
| N.º de referencia del gen | Q8NFD2 |
| Alias de gen | PKK2 |
| Mostrar más |
For Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.