missing translation for 'onlineSavingsMsg'
Learn More

RAB38 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Código de producto. 16899823 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
20 μL
150 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16899823 20 μL 20 microlitros
16889823 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16899823 Supplier Proteintech Supplier No. 122341AP20UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

May be involved in melanosomal transport and docking. Involved in the proper sorting of TYRP1.

Specifications

Antígeno RAB38
Aplicaciones Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Clasificación Polyclonal
Concentración 0.33 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica RAB38
N.º de referencia del gen P57729, Q8QZZ8
Alias de gen Melanoma antigen NY MEL 1, NY MEL 1, RAB38, Ras related protein Rab 38, rrGTPbp
Símbolos de los genes Rab38
Especie del huésped Rabbit
Inmunógeno RAB38 Fusion Protein Ag2874
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 23682, 72433
Especies diana Mouse, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.