Learn More
Veolia Water Systems R7 Reverse Osmosis System
Código de producto. 11964877
Cambiar vista
Click to view available options
Flow Rate: 7.5L/hour
Código de producto.
11964877 Flow Rate: 7.5L/hour
1 options
Código de producto. 11964877 Proveedor Veolia Water Systems N.º de proveedor OR007XXM1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos
Rich, robust, reliable purification system features osmosis with demineralization.
  • Purification of water by reverse osmosis
  • Purification system option R 7L/H
  • Pretreatment and polishing systems
  • Osmosis with demineralization
Especificaciones
Caudal 7.5 L/hr.