Learn More
Tocris Bioscience™ VU 0469650 Hydrochloride
Descripción
VU 0469650 hydrochloride is a potent and selective negative allosteric modulator of mGlu1 (IC50 = 99 nM). Exhibits >100-fold selectivity for mGlu1 over mGlu2-8 and 68 other GPCRs, ion channels, kinases and transporters. Brain penetrant.
Especificaciones
Especificaciones
| Nombre del producto químico o material | VU 0469650 hydrochloride |
| CAS | 1443748-47-7 |
| Sinónimo | 3-[(3R)-3-Methyl-4-(tricyclo[3.3.1.13,7]dec-1-ylcarbonyl)-1-piperazinyl]-2-pyridinecarbonitrile hydrochloride |
| Cantidad | 10 mg |
| Solubilidad | Soluble to 100mM in DMSO and to 50mM in ethanol |
| Fórmula molecular | C22H28N4O·HCl |
| Formula Weight (peso de la fórmula) | 400.94 |
| Forma física | Sólido |
| Tipo de producto | Bioactive Small Molecule |
| Pureza | >98% |
Research Use Only. Not For Human or Veterinary Use