Learn More
Tocris Bioscience™ KRIBB11
Descripción
KRIBB11 is a heat shock factor (HSF) inhibitor (IC50 = 1.2 μM). Increases apoptosis in cancer cells treated with the HSP90 inhibitors Geldanamycin (Cat. No. 1368) and 17-AAG (Cat. No. 1515). Blocks downstream induction of HSP27 and HSP70. Inhibits tumor growth in vivo.
Especificaciones
Especificaciones
| Nombre del producto químico o material | KRIBB11 |
| CAS | 342639-96-7 |
| Sinónimo | N2-1H-Indazol-5-yl-N6-methyl-3-nitro-2,6-pyridinediamine |
| Cantidad | 10 mg |
| Solubilidad | Soluble to 100mM in DMSO |
| Fórmula molecular | C13H12N6O2 |
| Formula Weight (peso de la fórmula) | 284.27 |
| Forma física | Sólido |
| Tipo de producto | Bioactive Small Molecule |
| Pureza | >99% |
Research Use Only. Not For Human or Veterinary Use