Learn More
Tocris Bioscience™ Edaglitazone
Descripción
Edaglitazone is a potent and selective PPARγ agonist (EC50 values are 35.6 and 1053 nM for PPARγ and PPARα cofactor recruitment respectively. Enhances insulin sensitivity in obese, but not lean, rats. Antidiabetic and orally bioavailable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Edaglitazone |
| CAS | 213411-83-7 |
| Sinónimo | 5-[[4-[2-(5-Methyl-2-phenyl-4-oxazolyl)ethoxy]benzo[b]thien-7-yl]methyl]-2,4-thiazolidinedione |
| Cantidad | 10 mg |
| Solubilidad | Soluble to 100mM in DMSO |
| Fórmula molecular | C24H20N2O4S2 |
| Formula Weight (peso de la fórmula) | 464.56 |
| Forma física | Sólido |
| Tipo de producto | Bioactive Small Molecule |
| Pureza | >98% |
Research Use Only. Not For Human or Veterinary Use