Learn More
Tocris Bioscience™ Deltarasin
Descripción
Deltarasin is a high affinity PDEδ-KRAS interaction inhibitor (Kd = 41 nM); binds to PDEδ. Reduces proliferation and induces cell death of KRAS-dependent Panc-Tu-1 and Capan-1 pancreatic cancer cells. Attenuates tumor growth of Panc-Tu-1 xenografts in mice.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Other Ras GTPase Compounds |
| CAS | 1440898-82-7 |
| Sinónimo | 2-[4-[(2S)-2-(2-Phenyl-1H-benzimidazol-1-yl)-2-(4-piperidinyl)ethoxy]phenyl]-1-(phenylmethyl)-1H-benzimidazole trihydrochloride |
| Cantidad | 10 mg |
| Solubilidad | Soluble to 100mM in water and to 100mM in DMSO |
| Fórmula molecular | C40H37N5O·3HCl |
| Formula Weight (peso de la fórmula) | 713.14 |
| Forma física | Sólido |
| Tipo de producto | Bioactive Small Molecule |
| Pureza | >99% |
Research Use Only. Not For Human or Veterinary Use