Learn More
Tocris Bioscience™ AM 6545
Descripción
AM 6545 is a high affinity and selective CB1 antagonist (Ki = 3.3 nM). Exhibits >100-fold selectivity for CB1 over CB2 receptors. Improves glucose homeostasis, reverses hepatic steatosis and reduces body weight in diet-induced obese mice. Orally available and non-brain penetrant.
Especificaciones
Especificaciones
| Nombre del producto químico o material | AM 6545 |
| CAS | 1245626-05-4 |
| Sinónimo | 5-[4-(4-Cyano-1-butyn-1-yl)phenyl]-1-(2,4-dichlorophenyl)-N-(1,1-dioxido-4-thiomorpholinyl)-4-methyl-1H-pyrazole-3-carboxamide |
| Cantidad | 10 mg |
| Solubilidad | Soluble to 20mM in DMSO |
| Fórmula molecular | C26H23Cl2N5O3S |
| Formula Weight (peso de la fórmula) | 556.46 |
| Forma física | Sólido |
| Tipo de producto | Bioactive Small Molecule |
| Pureza | >98% |
Research Use Only. Not For Human or Veterinary Use