Learn More
(R)-BAY1238097, MedChemExpress
Código de producto. 16494618
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16494618 10 mg 10 miligramos
16480917 50 mg 50 miligramos
16457337 5 mg 5 miligramos
16455466 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16494618 Proveedor Medchem Express N.º de proveedor HY112316A
MedChemExpress (R)-BAY1238097 is the R-isomer with lower activity of BAY1238097. BAY1238097 is a potent and selective inhibitor of BET binding to histones and has strong anti-proliferative activity in different AML (acute myeloid leukemia) and MM (multiple myeloma) models through down-regulation of c-Myc levels and its downstream transcriptome.
(R)-BAY1238097 is the R-isomer with lower activity of BAY1238097. BAY1238097 is a potent and selective inhibitor of BET binding to histones and has strong anti-proliferative activity in different AML (acute myeloid leukemia) and MM (multiple myeloma) models through down-regulation of c-Myc levels and its downstream transcriptome.
Especificaciones
Nombre del producto químico o material (R)-BAY1238097
CAS 1564269-85-7
Color Light Yellow
Notas de grado de pureza Research
Fórmula molecular C25H33N5O3
Cantidad 10 mg
Información de solubilidad DMSO : 150 mg/mL (332.18 mM; Need ultrasonic)
SMILES O=C(N1N=C(C2=CC=C(N3CCN(C)CC3)C=C2)C4=CC(OC)=C(OC)C=C4C[C@H]1C)NC
Peso molecular (g/mol) 451.56
Formula Weight (peso de la fórmula) 451.56
Porcentaje de pureza 98.2%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Neuroscience-Neuromodulation