Learn More
Ácido (R)-(-)-2-hidroxi-2-fenilpropiónico, + 98 %, Thermo Scientific Chemicals
Código de producto. 11392626
Cambiar vista
Click to view available options
Cantidad:
250 mg
1 g
Tamaño de la unidad:
1 g
250 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
11392626 1 g 1 g
11302636 250 mg 250 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 11392626 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor L08644.03
Solicitar formato a granel o personalizado
It is used in the asymmetric synthesis. Part III. Stereospecific synthesis of (R)-2-hydroxy-2-phenylpropionic acid, and (R)- and (S)-2-cyclohexyl-2-hydroxy-2-phenylacetic acid. Configurational relationship between (R)(-)-2-hydroxy-2-phenylpropionic acid and (S)(+)-2-phenylpropionic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
Se utiliza en la síntesis asimétrica. Part III. Stereospecific synthesis of (R)-2-hydroxy-2-phenylpropionic acid, and (R)- and (S)-2-cyclohexyl-2-hydroxy-2-phenylacetic acid. Configurational relationship between (R)(-)-2-hydroxy-2-phenylpropionic acid and (S)(+)-2-phenylpropionic acid.
Notas
Almacenar alejado de agentes oxidantes. Mantener el recipiente bien cerrado en un lugar fresco, seco y bien ventilado.
TRUSTED_SUSTAINABILITY
CAS 3966-30-1
Fórmula molecular C9H10O3
Peso molecular (g/mol) 166.176
Número MDL MFCD00067699
Clave InChI NWCHELUCVWSRRS-SECBINFHSA-N
Sinónimo r---2-hydroxy-2-phenylpropionic acid, 2r-2-hydroxy-2-phenylpropanoic acid, unii-h38dkr9931, r-2-hydroxy-2-phenylpropanoic acid, phenyllactic acid, atrolactic acid,-, 2-hydroxy-2-phenylpropanoic acid #, r---atrolactic acid, r-phenyl lactic acid, r-alpha-hydroxy-alpha-methylbenzeneacetic acid
PubChem CID 736858
Nombre IUPAC ácido (2R)-2-hidroxi-2-fenilpropanoico
SMILES CC(C1=CC=CC=C1)(C(=O)O)O
Especificaciones
Nombre del producto químico o material (R)-(-)-2-Hydroxy-2-phenylpropionic acid
Punto de fusión 115°C to 117°C
Cantidad 1 g
Beilstein 2936598
Rotación óptica −49° (c=2 in Water)
Formula Weight (peso de la fórmula) 166.18
Porcentaje de pureza ≥98%