Learn More
Tocris Bioscience™ QWF
Descripción
QWF is a tripeptide substance P (SP) antagonist (IC50 = 90 μM). Also inhibits binding of SP to Mas-related GPCR (MRGPR) X2. Inhibits SP-induced IgE-independent degranulation of mast cells in vitro. Inhibits compound 48/80-induced MRGPRX2 activation and scratching in mice in vivo.
Especificaciones
Especificaciones
| Pureza | 90% |
| CAS | 126088-82-2 |
| Contenido y almacenamiento | Almacene a -20 °C. |
| Cantidad | 5 mg |