Learn More
Quantikine Immunoassay Control Set 887 Human TIM-3, R&D Systems™
Código de producto. 18748998
Change view
Click to view available options
Cantidad:
1 Each
Tamaño de la unidad:
cada uno
18748998 1 Each cada uno
1 options
Código de producto. 18748998
Marca: R&D Systems QC217
For use with catalog number DTIM30
TIM-3 specifically detects TIM-3 in Human samples. It is validated for Control.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno TIM-3
Dilución Control
Alias de gen CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3
Cantidad 1 Each
Estado normativo RUO
ID de gen (Entrez) 84868
Reconstitución Reconstitute each vial with the volume of deionized or distilled water indicated on the product datasheet.
Especies diana Human
Contenido y almacenamiento Store the unopened product at 2 - 8 °C. Do not use past expiration date.