Learn More
R&D Systems™ Quantikine Immunoassay Control Group 235
Descripción
For use as quantitative controls for the determination of cytokine concentrations in biological fluids. Concentrations have been assigned using R&D Systems™’ Quantikine™ ELISA kits. Controls are prepared in a buffered protein base with preservatives. They contain recombinant human cytokines at low, medium and high concentrations. Controls are supplied lyophilized.
Especificaciones
Especificaciones
| Contenido y almacenamiento | Store the unopened product at 2-8°C. Do not use past expiration date. |
| Tipo | Immunoassay Control |
| Línea de productos | R&D Systems |
| Cantidad | 1 unidad |