Learn More
R&D Systems™ Quantikine Control Set 996 for Human Pentraxin 3, DPTX30B
Descripción
Quantikine Control Set 996 is designed for use with the human Pentraxin 3 (PTX3) Quantikine ELISA kit (catalog number DPTX30B).
- Species Reactivity: Human
- Application: ELISA control
- Components: Contains ProClin 300
- Storage: Store unopened product at 2°C to 8°C
- Usage: Research use only
Especificaciones
Especificaciones
| Analitos detectables | Pentraxin 3 (TSG-14) |
| Tipo | Immunoassay Control |
| Línea de productos | R&D Systems |
| Cantidad | 1 unidad |