Learn More
Tocris Bioscience™ QO 58
Description
QO 58 is a Kv7 channel opener (EC50 values are 0.6, 1.0, 5.2 and 7.0 μM for Kv7.4, Kv7.2, Kv7.3/7.5 and Kv7.1, respectively). Increases the pain threshold of neuropathic pain in a sciatic nerve CCI in vivo model.
Specifications
Specifications
| Nombre del producto químico o material | 5-(2,6-Dichloro-5-fluoro-3-pyridinyl)-3-phenyl-2-(trifluoromethyl)-pyrazolo[1,5-a]pyrimidin-7(4H)-one |
| CAS | 1259536-62-3 |
| Cantidad | 50 mg |
| Objetivo | Voltage-gated Potassium Channel Activators |
| Fórmula molecular | C18H8Cl2F4N4O |
| Formula Weight (peso de la fórmula) | 443.18 |
| Pureza | >98% |