Learn More
Tocris Bioscience™ QO 58
GREENER_CHOICE
Código de producto. 18702158
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18702158 10 mg 10 miligramos
18710939 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18702158 Proveedor Tocris Bioscience™ N.º de proveedor 5083/10
Kv7 channel opener
QO 58 is a Kv7 channel opener (EC50 values are 0.6, 1.0, 5.2 and 7.0 μM for Kv7.4, Kv7.2, Kv7.3/7.5 and Kv7.1, respectively). Increases the pain threshold of neuropathic pain in a sciatic nerve CCI in vivo model.
TRUSTED_SUSTAINABILITY
Especificaciones
Nombre del producto químico o material 5-(2,6-Dichloro-5-fluoro-3-pyridinyl)-3-phenyl-2-(trifluoromethyl)-pyrazolo[1,5-a]pyrimidin-7(4H)-one
CAS 1259536-62-3
Cantidad 10 mg
Objetivo Voltage-gated Potassium Channel Activators
Fórmula molecular C18H8Cl2F4N4O
Formula Weight (peso de la fórmula) 443.18
Pureza >98%