Learn More
Tocris Bioscience™ PZ 3022
Descripción
PZ 3022 is a potent allosteric pantothenate kinase (PanK) activator (EC50=5.3 nM for PanK3). In a mouse model of propionic acidemia, treatment with PZ 3022 increases hepatic CoASH and C2-CoA, and decreases C3-CoA, improving intracellular C3:C2-CoA ratio and plasma C3:C2-carnitine ratios. PZ 3022 is orally bioavailable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 6-[4-[2-(4-Cyclopropylphenyl)acetyl]-1-piperazinyl]-3-pyridazinecarbonitrile |
| CAS | 2170608-85-0 |
| Cantidad | 10 mg |
| Objetivo | Enzyme Activators |
| Fórmula molecular | C20H21N5O |
| Pureza | 0.98 |