Learn More
Mezcla de alcaloides de pirrolizidina 661 100 μg/mL en metanol, Dr. Ehrenstorfer
Especificaciones
Especificaciones
| Nombre del producto químico o material | (+)-Echimidine N-Oxide∣Echimidine∣Europine∣Europine-N-oxide∣Heliotrine∣Heliotrine-N-oxide∣Intermedine∣Intermedine N-oxide∣Lasiocarpine∣Lasiocarpine-N-oxide∣Lycopsamine∣Lycopsamine N-oxide∣Retrorsine∣Retrorsine N-Oxide∣Senecionine∣Senecionine N-Oxide∣Seneciphyllin∣Seneciphylline N-Oxide∣Senecivernine∣Senecivernine-N-oxide∣Senkirkine |
| CAS | 41093-89-4, 520-68-3, 570-19-4, 65582-53-8, 303-33-3, 6209-65-0 |
| Nombres de analitos o componentes | (+)-Echimidine N-Oxide∣Echimidine∣Europine∣Europine-N-oxide∣Heliotrine∣Heliotrine-N-oxide∣Intermedine∣Intermedine N-oxide∣Lasiocarpine∣Lasiocarpine-N-oxide∣Lycopsamine∣Lycopsamine N-oxide∣Retrorsine∣Retrorsine N-Oxide∣Senecionine∣Senecionine N-Oxide∣Seneciphyllin∣Seneciphylline N-Oxide∣Senecivernine∣Senecivernine-N-oxide∣Senkirkine |
| Condiciones de envío | +20°C |
| Certificaciones/conformidad | ISO 17034 |
| Solvent or Matrix | Methanol |
| Cantidad | 1 mL |
| Concentration or Composition (by Analyte or Components) | (+)-Echimidine N-Oxide 100 μg/mL∣Echimidine 100 μg/mL∣Europine 100 μg/mL∣Europine-N-oxide 100 μg/mL∣Heliotrine 100 μg/mL∣Heliotrine-N-oxide 100 μg/mL∣Intermedine 100 μg/mL∣Intermedine N-oxide 100 μg/mL∣Lasiocarpine 100 μg/mL∣Lasiocarpine-N-oxide 100 μg/mL∣Lycopsamine 100 μg/mL∣Lycopsamine N-oxide 100 μg/mL∣Retrorsine 100 μg/mL∣Retrorsine N-Oxide 100 μg/mL∣Senecionine 100 μg/mL∣Senecionine N-Oxide 100 μg/mL∣Seneciphyllin 100 μg/mL∣Seneciphylline N-Oxide 100 μg/mL∣Senecivernine 100 μg/mL∣Senecivernine-N-oxide 100 μg/mL∣Senkirkine 100 μg/mL |