Learn More
Platos de cristalización de vidrio de borosilicato Pyrex™
Reusable borosilicate glass dishes with heavy duty rims ideal for storage and crystallization.
Marca:  Pyrex 1470/10D
Código de producto. 10652291
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos »
Descripción
Descripción
  • Complies to DIN 12337
  • Manufactured from Pyrex™ borosilicate glass
  • Suitable for use up to 400°C
  • Chemically and thermally resistant
TRUSTED_SUSTAINABILITY
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones