missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSGR Polyclonal Antibody, Invitrogen™
Descripción
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Galago, Marmoset, Hamster, Bat, Rabbit, Pig (100%).
PSGR (prostate-specific G-protein coupled receptor), also known as OR51E2 (olfactory receptor 51E2), is a 320 amino acid multi-pass membrane protein that belongs to the olfactory receptor subfamily of G-protein coupled receptors. Expressed exclusively in prostate tissue and upregulated in prostate cancer, PSGR functions as an odorant receptor that binds odorant molecules and triggers the perception of smell.
Especificaciones
Especificaciones
| Antígeno | PSGR |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.1% sodium azide |
| génica | OR51E2 |
| N.º de referencia del gen | Q9H255 |
| Alias de gen | 4633402A21Rik; GA_x6K02T2PBJ9-5459657-5458695; G-protein coupled receptor RA1c; hCG_2044604; HPRAJ; Mol2.3; MOR18-2; olfactory receptor 51E2; Olfactory receptor 59; olfactory receptor 78; olfactory receptor family 51 subfamily E member 2; olfactory receptor MOR18-2; olfactory receptor OR11-16; olfactory receptor, family 51, subfamily E, member 2; olfactory receptor, family 51, subfamily E, member 3 pseudogene; olfactory receptor, family 52, subfamily A, member 2; Olfr78; Olr59; OR51E2; OR51E3P; OR52A2; prostate specific G-protein coupled receptor; prostate-specific G-protein coupled receptor; PSGR; RA1c |
| Símbolos de los genes | OR51E2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?