Learn More
Pseudoginsenoside F11, MedChemExpress
Código de producto. 16409018
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16409018 10 mg 10 miligramos
16401438 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16409018 Proveedor Medchem Express N.º de proveedor HYN0541
MedChemExpress Pseudoginsenoside F11 (Ginsenoside A1), a component of Panax quinquefolium (American ginseng), has been demonstrated to antagonize the learning and memory deficits induced by scopolamine, morphine and methamphetamine in mice.
Pseudoginsenoside F11 (Ginsenoside A1), a component of Panax quinquefolium (American ginseng), has been demonstrated to antagonize the learning and memory deficits induced by scopolamine, morphine and methamphetamine in mice.
Especificaciones
Nombre del producto químico o material Pseudoginsenoside F11
CAS 69884-00-0
Color Light Yellow
Notas de grado de pureza Research
Fórmula molecular C42H72O14
Cantidad 10 mg
Sinónimo Ginsenoside A1
Información de solubilidad DMSO : 100 mg/mL (124.84 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (0.84 mM; Need ultrasonic)
SMILES C[C@@]([C@@]12C)(C[C@H](O[C@@](O[C@H](CO)[C@@H](O)[C@@H]3O)([H])[C@@H]3O[C@@](O[C@@H](C)[C@H](O)[C@H]4O)([H])[C@@H]4O)[C@@]5([H])C6(C)C)[C@@](C[C@@H](O)[C@]1([H])[C@]([C@]7(O[C@@H](C(C)(O)C)CC7)C)([H])CC2)([H])[C@]5(CC[C@@H]6O)C
Peso molecular (g/mol) 801.01
Formula Weight (peso de la fórmula) 801.01
Porcentaje de pureza 98.0%
Grado Research
Forma física Solid