missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRSS27 Monoclonal Antibody (06)
GREENER_CHOICE

Código de producto. 16314284
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16314284 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16314284 Proveedor Invitrogen™ N.º de proveedor MA530788

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Pancreasin/Marapsin/PRSS27.

PRSS27, also known as Marapsin, is a gene located on chromosome 16p13.3 that encodes a serine protease enzyme. This protein is part of the trypsin-like serine protease family, which is characterized by a catalytic triad essential for proteolytic activity. PRSS27 is primarily expressed in the epidermis and other epithelial tissues, such as the salivary glands and pancreas. It plays a role in various physiological processes, including digestion, inflammation, and immune response regulation. In the skin, PRSS27 is involved in keratinocyte differentiation and the maintenance of skin barrier function. Dysregulation or altered expression of PRSS27 has been associated with inflammatory skin conditions, such as psoriasis, where it may influence the inflammatory cascade and contribute to disease pathogenesis. Research into PRSS27 continues to explore its potential role in epithelial homeostasis and as a therapeutic target for skin-related and other inflammatory disorders.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno PRSS27
Aplicaciones ELISA
Clasificación Monoclonal
Clon 06
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative
génica PRSS27
N.º de referencia del gen Q9BQR3
Alias de gen CAP-2; CAPH2; channel-activating protease 2; channel-activating protease 2-like protein; EC 3.4.21; EC 3.4.21.4; EC 3.4.21.43; marapsin; Mpn; MPNMarapsin; Pancreasin; protease, serine 27; PRSS27; Serine protease 27; UNQ1884/PRO4327
Símbolos de los genes PRSS27
Especie del huésped Mouse
Inmunógeno Recombinant Human Pancreasin/Marapsin/PRSS27 protein (Met1-Lys290).
Método de purificación Protein A
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 83886
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.