missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRSS21 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15538372
Cambiar vista
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15538372 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15538372 Proveedor Invitrogen™ N.º de proveedor PA547879

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Goat Polyclonal Antibody

This antibody detects recombinant mouse Testisin/Prss21 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) Prss8 and rmPrss27 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

PRSS21, also known as testisin, is a serine protease gene located on human chromosome 16p13.3. This gene is primarily expressed in the testes, where it plays a significant role in spermatogenesis, particularly during the development and maturation of sperm cells. Structurally, PRSS21 encodes a glycosylphosphatidylinositol (GPI)-anchored serine protease, a feature that helps localize it to the cell surface, facilitating its involvement in cell signaling and proteolytic pathways. While research into PRSS21 remains ongoing, its specific function is believed to be related to remodeling the extracellular matrix during sperm development, impacting fertility. Beyond reproductive roles, alterations in PRSS21 expression have been observed in some cancer types, such as testicular germ cell tumors, suggesting its potential role as a biomarker for certain malignancies. This dual functionality underscores its importance not only in reproductive biology but also within oncology research.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno PRSS21
Aplicaciones Immunohistochemistry (Frozen), Western Blot
Clasificación Polyclonal
Concentración 0.2 mg/mL
Conjugado Unconjugated
Formulación PBS with 5% trehalose and No Preservative
génica Prss21
N.º de referencia del gen Q9JHJ7
Alias de gen 1700023E12Rik; EC 3.4.21; eosinophil serine protease 1; eosinophil serine protease-1; ESP1; ESP-1; mT4; protease, serine 21; protease, serine, 21; protease, serine, 21 (testisin); PRSS21; Resp-1; Serine protease 21; serine protease from eosinophils; TESP5; TEST1; TEST1testisin; testicular serine protease 5; testisin; Tryptase 4; UNQ266/PRO303
Símbolos de los genes Prss21
Especie del huésped Goat
Inmunógeno Mouse myeloma cell line NS0-derived recombinant mouse Testisin/Prss21 Gln28-Asn298.
Método de purificación Antigen affinity chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 353251, 57256
Especies diana Mouse, Rat
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.