missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Protocadherin Gamma Monoclonal Antibody (N159/5)
GREENER_CHOICE

Código de producto. 16312134
Cambiar vista
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16312134 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16312134 Proveedor Invitrogen™ N.º de proveedor MA527615
 Artículo restringido
Fecha estimada de envoi: 11-08-2026
Añadir a la cesta
Añadir a la cesta
Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

1 μg/mL of MA5-27615 was sufficient for detection of Protocadherin gamma (pan) in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 100kDa. Cross-reacts with all Gamma-protocadherin-A, -B and -C proteins. This antibody was formerly sold as clone S159-5.

The protocadherin gamma gene cluster is one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Protocadherin Gamma
Aplicaciones Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon N159/5
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
génica PCDHGA12
N.º de referencia del gen O60330, Q91XY4, Q9UN70, Q9UN71, Q9Y5F6, Q9Y5F7, Q9Y5F8, Q9Y5F9, Q9Y5G0, Q9Y5G1, Q9Y5G2, Q9Y5G3, Q9Y5G4, Q9Y5G5, Q9Y5G6, Q9Y5G7, Q9Y5G8, Q9Y5G9, Q9Y5H0, Q9Y5H1, Q9Y5H2, Q9Y5H3, Q9Y5H4
Alias de gen 3' RACE product; cadherin 20; cadherin 21; cadherin ME3; cadherin ME6; cadherin-20; Cadherin-21; cadherin-like 2; CDH20; CDH21; CDH-GAMMA-A5; FIB2; FIB3; fibroblast cadherin FIB2; fibroblast cadherin FIB3; fibroblast cadherin-2; Fibroblast cadherin-3; KIAA0327; KIAA0588; ME3; ME6; mKIAA0588; mKIAA4054; Pan-Gamma-Protocadherin-A; pc2c; PC43; PC-43; PCDH gamma A; Pcdh13; PCDH2; PCDHGA; PCDHGA1; PCDHGA10; PCDHGA11; PCDHGA12; Pcdhga2; PCDHGA3; Pcdhga4; PCDHGA5; PCDHGA6; PCDHGA7; Pcdhga8; PCDHGA9; PCDH-gamma-A; PCDH-GAMMA-A1; PCDH-gamma-A10; PCDH-GAMMA-A11; PCDH-gamma-A12; PCDH-GAMMA-A2; PCDH-gamma-A3; PCDH-gamma-A4; PCDH-GAMMA-A5; PCDH-gamma-A6; PCDH-GAMMA-A7; PCDH-gamma-A8; PCDH-gamma-A9; PCDH-GAMMA-B1; PCDH-GAMMA-B2; PCDH-gamma-B3; PCDH-GAMMA-B4; PCDH-GAMMA-B5; PCDH-GAMMA-B6; PCDH-gamma-B7; PCDH-GAMMA-C3; PCDH-gamma-C4; PCDH-gamma-C5; Pcdhgb1; PCDHGB2; PCDHGB3; PCDHGB4; PCDHGB5; Pcdhgb6; Pcdhgb7; PCDHGC3; Pcdhgc4; PCDHGC5; protocadherin 13; protocadherin 2; protocadherin 43; protocadherin gamma A12; protocadherin gamma B2; protocadherin gamma B6; protocadherin gamma C-VI; protocadherin gamma subfamily A, 1; protocadherin gamma subfamily A, 10; protocadherin gamma subfamily A, 11; protocadherin gamma subfamily A, 12; protocadherin gamma subfamily A, 2; protocadherin gamma subfamily A, 3; protocadherin gamma subfamily A, 4; protocadherin gamma subfamily A, 5; protocadherin gamma subfamily A, 6; protocadherin gamma subfamily A, 7; protocadherin gamma subfamily A, 8; protocadherin gamma subfamily A, 9; protocadherin gamma subfamily B, 1; protocadherin gamma subfamily B, 1, isoform 2; protocadherin gamma subfamily B, 2; protocadherin gamma subfamily B, 3; protocadherin gamma subfamily B, 4; protocadherin gamma subfamily B, 5; protocadherin gamma subfamily B, 6; protocadherin gamma subfamily B, 7; protocadherin gamma subfamily C, 3; protocadherin gamma subfamily C, 4; protocadherin gamma subfamily C, 5; protocadherin gamma-A1; Protocadherin gamma-A10; protocadherin gamma-A11; Protocadherin gamma-A12; protocadherin gamma-A2; Protocadherin gamma-A3; protocadherin gamma-A4; Protocadherin gamma-A5; protocadherin gamma-A6; Protocadherin gamma-A7; protocadherin gamma-A8; Protocadherin gamma-A9; protocadherin gamma-B1; Protocadherin gamma-B2; protocadherin gamma-B3; Protocadherin gamma-B4; protocadherin gamma-B5; Protocadherin gamma-B6; protocadherin gamma-B7; Protocadherin gamma-C3; protocadherin gamma-C4; Protocadherin gamma-C5; protocadherin-2; Protocadherin-43; UNQ371/PRO707
Símbolos de los genes Pcdhga1, Pcdhga10, Pcdhga11, PCDHGA12, Pcdhga2, PCDHGA3, PCDHGA4, Pcdhga5, PCDHGA6, Pcdhga7, PCDHGA8, Pcdhga9, PCDHGB1, PCDHGB2, PCDHGB3, Pcdhgb4, Pcdhgb5, Pcdhgb6, Pcdhgb7, Pcdhgc3, PCDHGC4, PCDHGC5
Especie del huésped Mouse
Inmunógeno Fusion protein corresponding to amino acids 808-931 (C-terminal cytoplasmic constant domain) of mouse Protocadherin-gamma-A1 that is shared by all 22 Gamma-protocadherins (A subfamily amino acids ~807-930, B subfamily amino acids ~789-912 and C subfamily amino acids ~818-941). 99% identity with human (123/124 amino acids).
Método de purificación Protein G
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 102552497, 102552705, 102552796, 116782, 252894, 252895, 252897, 26025, 291635, 291637, 364843, 364844, 498846, 498847, 498849, 498850, 5098, 553129, 56097, 56098, 56099, 56100, 56101, 56102, 56103, 56104, 56105, 56106, 56107, 56108, 56109, 56110, 56111, 56112, 56113, 56114, 641525, 641526, 680361, 680448, 8641, 93699, 93700, 93701, 93702, 93703, 93704, 93706, 93707, 93708, 93709, 93710, 93711, 93712, 93713, 93714, 93715, 93716, 93717, 93722, 93723, 93724, 9708
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.