Learn More
Protein kinase inhibitors 1, MedChemExpress
Código de producto. 16444106
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
Tamaño de la unidad:
10 miligramos
25 miligramos
5 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16444106 25 mg 25 miligramos
16432648 5 mg 5 miligramos
16420449 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16444106 Proveedor Medchem Express N.º de proveedor HYU00439
MedChemExpress Protein kinase inhibitors 1 is a novel inhibitor of HIPK2 with an IC50 of 74 nM and Kd of 9.5 nM.
Protein kinase inhibitors 1 is a novel inhibitor of HIPK2 with an IC50 of 74 nM and Kd of 9.5 nM.
Especificaciones
Nombre del producto químico o material Protein kinase inhibitors 1
CAS 1365986-44-2
Color Light Yellow
Notas de grado de pureza Research
Fórmula molecular C18H17N5O3S
Cantidad 25 mg
SMILES O=C(NC/1=O)SC1=C\C2=CNC(C(C3=CC=C(N4CCNCC4)N=C3)=C2)=O
Peso molecular (g/mol) 383.42
Formula Weight (peso de la fórmula) 383.42
Porcentaje de pureza 99.0%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease