Learn More
Tocris Bioscience™ Propidium iodide
Red-fluorescent DNA stain; membrane impermeant to live cells. Pi staining differentiates live and dead cells
Marca: Tocris Bioscience™ 5135/10
Descripción
Propidium iodide is a red-fluorescent DNA stain. Membrane impermeant to live cells. Propidium iodide staining differentiates between live and dead cells in cell populations.Especificaciones
| 95% | |
| 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide | |
| -20°C |
| Fluorescent DNA Stains | |
| 25535-16-4 | |
| 10 mg |
For Research Use Only