Learn More
Thermo Scientific Chemicals Clorhidrato de propafenona
Código de producto. 15841062 Tienda Thermo Scientific Chemicals Productos
Cambiar vista
Click to view available options
Cantidad:
1 g
5 g
Tamaño de la unidad:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
15841062 5 g 5 g
15811062 1 g 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 15841062 Proveedor Thermo Scientific Chemicals N.º de proveedor 462390050
Solicitar formato a granel o personalizado
CAS: 34183-22-7 | C21H28ClNO3 | 377.91 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS 34183-22-7
Fórmula molecular C21H28ClNO3
Peso molecular (g/mol) 377.91
Clave InChI XWIHRGFIPXWGEF-UHFFFAOYNA-N
Nombre IUPAC cloruro de hidrógeno 1-{2-[2-hidroxi-3-(propilamino)propoxi]fenil}-3-fenilpropan-1-ona
SMILES [H+].[Cl-].CCCNCC(O)COC1=CC=CC=C1C(=O)CCC1=CC=CC=C1
Especificaciones
Nombre del producto químico o material Propafenone hydrochloride
Punto de fusión 173.0°C
Color Transparente, sin color
Cantidad 5 g
Información de solubilidad Solubility aq. soln.: .. Other solubilities: soluble in methanol, insoluble in ether
Formula Weight (peso de la fórmula) 377.9