missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ JAK2[pY1007/1008] Human ProcartaPlex™ Simplex Kit

Código de producto. 30825497
Cambiar vista
Click to view available options
Cantidad:
96 pruebas
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad
30825497 96 pruebas
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30825497 Proveedor Invitrogen™ N.º de proveedor EPX01097121PHO
 Artículo restringido
Fecha estimada de envoi: 19-10-2026
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

ProcartaPlex Simplex kits enable precise analysis of the protein of interest.

The ProcartaPlex Human JAK2[pY1007/1008] Simplex Kit measures the level of phosphorylated human JAK2[pY1007/1008] in cell lysates. It is designed to be combinable with other Simplex kits so you can create your own multiplex panel that utilizes Luminex xMAP technology for protein quantitation. When combining multiple Simplex kits (i.e., when you are not using a pre-configured Multiplex Panel), only one buffer kit (sold separately) is needed for each assay plate regardless of plex size.

For preparation of cell lysates, we recommend the use of either ProcartaPlex Cell Lysis Buffer (Cat. No. EPX-99999-000) or Cell Extraction Buffer (Cat. No. FNN0011) supplemented with 0.5 M EDTA and Halt Protease and Phosphatase Inhibitor Single–Use Cocktail 100X (Cat. No. 78440 or 78442). These buffers are not included with the kit. Depending on your cell system, other buffers can be suitable as well.

The ProcartaPlex Human JAK2[pY1007/1008] Simplex Kit is part of the ProcartaPlex Human JAK-STAT Pathway Panel (phospho), 6plex (Cat. No. EPX060-97124-PHO) which complements the ProcartaPlex Human JAK-STAT Pathway Panel (total), 6plex (Cat. No. EPX060-97024-901).

About ProcartaPlex Assays for the Luminex Platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

  • More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
  • Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g. Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine). Visit thermofisher.com/procartaplex for more information, including a comprehensive list of individual protein targets.

TRUSTED_SUSTAINABILITY

Especificaciones

Tipo de producto Simplex Kit
Cantidad 96 pruebas
Especie Human
Combinability Combinable
Tipo de esfera Magnetic
Región de cordón JAK2[pY1007/1008] [55]
Sensibilidad del ensayo As determined for Lot 1: 8.4 U/mL
Intervalo del ensayo As determined for Lot 1: 15 - 62 900 U/mL
Formato Simplex Kit
Para utilizar con (equipo) Luminex™ Instruments
génica JAK2
Alias de gen Janus Kinase 2
ID de gen (Entrez) 3717
Símbolo de gen JAK2
CV interensayo As determined for Lot 1: 7.5%
CV intraensayo As determined for Lot 1: 6.4%
N.º plex tamaño multiplex Simplex
Línea de productos ProcartaPlex
Tipo de muestra Cell lysates
Volumen de muestra Cell lysates (prediluted): 25 μL
ID. de UniProt O60674
Sinónimo Janus Kinase 2
Contenido y almacenamiento See Certificate of Analysis
Condiciones de envío Wet Ice
Research Area Signaling
Mostrar más Mostrar menos

For Research Use Only.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.