Learn More
Pregnenolone monosulfate, MedChemExpress
Descripción
Pregnenolone monosulfate acts as a signaling-specific inhibitor of cannabinoid CB1 receptor, inhibits the effects of tetrahydrocannabinol (THC) that are mediated by the CB1 receptors. Pregnenolone monosulfate can protect the brain from cannabis intoxication. Pregnenolone monosulfate is also a TRPM3 channel activator, and also can weakly activate TRPM1 channels.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Pregnenolone monosulfate |
| CAS | 1247-64-9 |
| Color | Blanco |
| Notas de grado de pureza | Research |
| Fórmula molecular | C21H32O5S |
| Cantidad | 50 mg |
| Sinónimo | 3β-Hydroxy-5-pregnen-20-one monosulfate |
| Información de solubilidad | DMSO : 62.5 mg/mL (157.61 mM; Need ultrasonic) |
| SMILES | CC([C@H]1CC[C@@]2([H])[C@]3([H])CC=C4C[C@@H](OS(=O)(O)=O)CC[C@]4(C)[C@@]3([H])CC[C@]12C)=O |
| Peso molecular (g/mol) | 396.54 |
| Mostrar más |