Learn More
Pralatrexate, MedChemExpress
Código de producto. 16490327
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16490327 50 mg 50 miligramos
16486687 5 mg 5 miligramos
16453978 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16490327 Proveedor Medchem Express N.º de proveedor HY10446
MedChemExpress Pralatrexate is an antifolate and is a potent dihydrofolate reductasean (DHFR) inhibitor with a Ki of 13.4 pM. Pralatrexate is a substrate for folylpolyglutamate synthetase with improved cellular uptake and retention. Pralatrexate has antitumor activities and has the potential for relapsed/refractory T-cell lymphoma treatment.
Pralatrexate is an antifolate and is a potent dihydrofolate reductasean (DHFR) inhibitor with a Ki of 13.4 pM. Pralatrexate is a substrate for folylpolyglutamate synthetase with improved cellular uptake and retention. Pralatrexate has antitumor activities and has the potential for relapsed/refractory T-cell lymphoma treatment.
Especificaciones
Nombre del producto químico o material Pralatrexate
Forma física Solid
CAS 146464-95-1
Color Yellow
Cantidad 50 mg
Notas de grado de pureza Research
Fórmula molecular C23H23N7O5
Para utilizar con (aplicación) Cancer-programmed cell death
Información de solubilidad DMSO : ≥ 50 mg/mL (104.72 mM)
SMILES O=C(CC[C@H](NC(C1=CC=C(C=C1)C(CC#C)CC2=NC3=C(N=C(N=C3N=C2)N)N)=O)C(O)=O)O
Peso molecular (g/mol) 477.47
Formula Weight (peso de la fórmula) 477.47
Porcentaje de pureza 95.0%