Learn More
Tocris Bioscience™ PR 619
Broad spectrum DUB inhibitor
Marca: Tocris Bioscience™ 4482/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PR 619 is a broad spectrum, reversible DUB inhibitor. Exhibits limited activity against other proteases. Induces accumulation of polyubiquitinated proteins, but has no direct inhibitory effect on the proteasome. Cytotoxic in HEK293T and colorectal cancer cells. Stabilizes microtubule network in oligodendroglial cells.Especificaciones
| PR 619 | |
| 2,6-Diamino-3,5-dithiocyanopyridine | |
| Soluble to 10mM in DMSO | |
| Bioactive Small Molecule |
| 2645-32-1 | |
| 50 mg | |
| Solid |
Título del producto
Research Use Only. Not For Human or Veterinary Use