Learn More
Tocris Bioscience™ PPDA
GREENER_CHOICE
Código de producto. 18054802
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18054802 10 mg 10 miligramos
18085752 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18054802 Proveedor Tocris Bioscience™ N.º de proveedor 2530/10
GluN2C/GluN2D-selective NMDA antagonist
Subtype-selective NMDA receptor antagonist that preferentially binds to GluN2C/GluN2D (formally NR2C/NR2D) containing receptors (Ki values are 0.096, 0.125, 0.31 and 0.55 μM for GluN2C, GluN2D, GluN2B and GluN2A subunits respectively).
Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
TRUSTED_SUSTAINABILITY
CAS 684283-16-7
Fórmula molecular C21H18N2O5
Especificaciones
Nombre del producto químico o material PPDA
Cantidad 10 mg
Formula Weight (peso de la fórmula) 378.38
Pureza >98%
Peso molecular 378.384