missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPAR alpha Polyclonal Antibody
GREENER_CHOICE

Código de producto. 11529100
Cambiar vista
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11529100 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11529100 Proveedor Invitrogen™ N.º de proveedor PA1822A

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

PA1-822A detects peroxisome proliferator activated receptor (PPAR) alpha from mouse samples. PA1-822A has been successfully used in Western blot, immunoprecipitation and immunofluoresence procedures. By Western blot, this antibody detects an ~52 kDa protein representing PPAR alpha from 3T3-L1 cell lysate. Immunofluoresence staining is observed predominantly in the nucleus as well as the cytoplasm. PA1-822A immunogen is a synthetic peptide corresponding to residues M(1) V D T E S P I C P L S P L E A D D(18) C of mouse PPAR alpha. A single amino acid substitution (I8 to L8) exists between the mouse and human protein. This immunizing peptide (Cat.# PEP-025) is available for use in neutralization and control experiments.

Peroxisome proliferators are non-genotoxic carcinogens which are purported to exert their effect on cells through their interaction with members of the nuclear hormone receptor family termed peroxisome proliferator activated receptors (PPAR's). Nuclear hormone receptors are ligand-dependent intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate ligand. Studies indicate that PPAR's are activated by peroxisome proliferators such as clofibric acid, nafenopin, and WY-14,643, as well as by some fatty acids. It has also been shown that PPAR's can induce transcription of acyl coenzyme A oxidase & cytochrome P450 (CYP450) A6 through interaction with specific response elements. PPAR, like several other nuclear hormone receptors, heterodimerizes with retinoid X receptor (RXR) alpha.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno PPAR alpha
Aplicaciones Immunoprecipitation, Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1mg/mL BSA and 0.05% sodium azide
génica PPARA
N.º de referencia del gen P23204, Q07869
Alias de gen 4933429D07Rik; AW742785; hPPAR; I79_022392; MGC2237; MGC2452; Nr1c1; nuclear hormone receptor; nuclear receptor; nuclear receptor subfamily 1 group C member 1; peroxisome proliferative activated receptor alpha; peroxisome proliferative activated receptor, alpha; peroxisome proliferative activated receptor, alpha isoform 1; peroxisome proliferator; peroxisome proliferator activated receptor; peroxisome proliferator activated receptor alpha; peroxisome proliferator-activated nuclear receptor alpha variant 3; peroxisome proliferator-activated receptor alpha; PPAR; PPAR ALPHA; Ppara; PPARALPHA; PPAR-alpha; proliferator-activated receptor alpha; transcription factor belonging to the nuclear receptor superfamily
Símbolos de los genes PPARA
Especie del huésped Rabbit
Inmunógeno Synthetic peptide corresponding to residues M(1) V D T E S P I C P L S P L E A D D(18) C of mouse PPAR alpha.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 19013, 5465
Especies diana Human, Mouse
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.