missing translation for 'onlineSavingsMsg'
Learn More

Potassium titanium oxide

Product Code. 11409756
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
Product Code. Quantity unitSize
11409756 500 g 500g
11499746 100 g 100g
11419756 2500 g 2500g
3 options
This item is not returnable. View return policy

Product Code. 11409756

Brand: Thermo Scientific Alfa Aesar B20263.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12030-97-6 | K2O3Ti | 174.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12030-97-6
Molecular Formula K2O3Ti
Molecular Weight (g/mol) 174.06
MDL Number MFCD00058803
InChI Key KEWYKJOOCFOYTD-UHFFFAOYSA-N
Synonym kaliumtitanat, titanate,dipotassium, potassiumtitanatewhisker, 2k.o3ti, titanate tio32-,dipotassium, dipotassium;dioxido oxo titanium
PubChem CID 45052161
SMILES [O--].[O--].[O--].[K+].[K+].[Ti+4]

Specifications

Melting Point 1615°C
Quantity 500 g
IUPAC Name dipotassium;dioxido(oxo)titanium
Odor Odorless
Chemical Name or Material Potassium titanium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.