Learn More
Thermo Scientific™ Potassium hydride, 30% w/w in mineral oil
Código de producto. 17980050
Cambiar vista
Click to view available options
Cantidad:
5 g
Código de producto. Cantidad
17980050 5 g
1 options
Código de producto. 17980050 Proveedor Thermo Scientific™ N.º de proveedor L13266.NB
 Artículo restringido
En stock
Añadir a la cesta
CAS: 7693-26-7 | HK | 40.11 g/mol
Potassium hydride is a precursor in the production of various bases by deprotonation, highly-hindered bases potassium triethylmethoxide and potassium 3-aminopropylamide It serves as a powerful base in organic synthesis of deprotonate carbonyl compounds and amines to form enolates and amides respectively.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Especificaciones
CAS 7693-26-7
Densidad 1.45
Cantidad 5 g
Temperatura de inflamación 160°(320°F)
Formulario Liquid
Fórmula molecular HK
Número MDL MFCD00011357
Número UN UN1409
Fieser 1,935 12,407 13,257 14,265 17,290 18,299 19,276 21,360
Clave InChI OCFVSFVLVRNXFJ-UHFFFAOYSA-N
SMILES [H-].[K+]
Nombre IUPAC potassium hydride
Peso molecular (g/mol) 40.11
Formula Weight (peso de la fórmula) 40.11