Learn More
Tocris Bioscience™ Pomalidomide 5'-piperazine-4-methylpiperidine
Descripción
Pomalidomide 5'-piperazine-4-methylpiperidine is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand with a terminal piperidine for onward chemistry to generate Protein Degraders with rigid linkers. Part of a range of functionalized tool molecules for PROTAC™. Please contact us for SD files of our available Degrader Building Blocks.PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 2-(2,6-Dioxo-3-piperidinyl)-5-[4-(4-piperidinylmethyl)-1-piperazinyl]-1H-isoindole-1,3(2H)-dione dihydrochloride |
| CAS | 2758431-93-3 |
| Cantidad | 25 mg |
| Objetivo | Degrader Building Blocks |
| Fórmula molecular | C23H29N5O4.2HCl |
| Pureza | 95% |