Learn More
Tocris Bioscience™ Pomalidomide 4'-PEG4-acid
Descripción
Pomalidomide 4'-PEG4-acid is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a PEG4 linker with terminal carboxylic acid ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 3-[2-[2-[2-[2-[[2-(2,6-Dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1 H-isoindol-4-yl]amino]ethoxy]ethoxy]ethoxy]ethoxy]propanoic acid |
| CAS | 2138440-81-8 |
| Cantidad | 25 mg |
| Objetivo | Degrader Building Blocks |
| Fórmula molecular | C24H31N3O10 |
| Pureza | 0.95 |