Learn More
Tocris Bioscience™ Pomalidomide 4'-PEG2-amine
Descripción
Pomalidomide 4'-PEG2-amine is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a PEG2 linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
This product has been recently renamed. The previous name for this product was Pomalidomide - linker 1
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4-[[2-[2-(2-Aminoethoxy)ethoxy]ethyl]amino]-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride |
| CAS | 2245697-87-2 |
| Sinónimo | Pomalidomide - linker 1 |
| Cantidad | 25 mg |
| Objetivo | Degrader Building Blocks |
| Fórmula molecular | C19H24N4O6.HCl |
| Formula Weight (peso de la fórmula) | 440.88 |
| Pureza | 95% |