Learn More
Tocris Bioscience™ PNU 142633
Descripción
Very selective, high affinity 5-HT1D receptor agonist (Ki values are 6 and > 18000 nM at human 5-HT1D and 5-HT1B receptors respectively). Inhibits sympathetically-induced tachycardic responses and blocks neurogenic plasma protein extravasation in vivo.
Especificaciones
Especificaciones
| Nombre del producto químico o material | PNU 142633 |
| CAS | 187665-65-2 |
| Cantidad | 10 mg |
| Fórmula molecular | C24H30N4O3 |
| Formula Weight (peso de la fórmula) | 422.53 |
| Pureza | >98% |
| Peso molecular | 422.529 |