missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLLP Polyclonal Antibody

Descripción
Positive test controls include: Mouse brain, Rat brain, Rat kidney. The target is usually found in the following locations: Membrane, Multi-pass membrane protein. Immunogen sequence: MAEFPSKVST RTSSPAQGAE ASVSALRPDL GFVRSRLGAL MLLQLVLGLL VWALIADTPY HLYPAYGWVM FVAVFLWLVT IVLFNLYLFQ LHMKLYMVPW.
Plasmolipin is an 18 kDa membrane-bound proteolipid that belongs to the tetraspan (4TM) family, a rapidly growing group of myelin-associated proteins. Many of these proteins could be linked to human hereditary demyelinating neuropathies. Studies indicate that plasmolipin is a component of the cytoskeletal structure and has been localized to the apical surface of tubular epithelial cells. It has been shown that, upon addition to lipid bilayers, plasmolipin induces formation of ion channels that are both voltage-dependent and potassium-specific. Plasmolipin was originally isolated from kidney tissue. Subsequently, it has been shown to be widely expressed, not only being present in kidney, but also in the nervous system, thymus, testis, lung, and thyroid gland.
Especificaciones
Especificaciones
| Antígeno | PLLP |
| Aplicaciones | ELISA, Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 1.15 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| génica | PLLP |
| N.º de referencia del gen | P47987, Q9DCU2 |
| Alias de gen | 0610010I06Rik; AV001002; Plapi; Plasma membrane proteolipid; plasma membrane proteolipid (plasmolipin); plasmolipin; PLLP; PMLP; Tm4sf11; transmembrane 4 superfamily member 11; transmembrane 4 superfamily member 11 (plasmolipin) |
| Símbolos de los genes | PLLP |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?