missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PITX3 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 10715703
Change view
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
50 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
10715703 50 μg 50 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10715703 Proveedor Invitrogen™ N.º de proveedor 382850

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

PITX3 Polyclonal Antibody for IHC (F), ICC/IF

This gene encodes a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. Members of this family act as transcription factors. This protein is involved in lens formation during eye development. Mutations of this gene have been associated with anterior segment mesenchymal dysgenesis and congenital cataracts.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno PITX3
Aplicaciones Immunohistochemistry (Frozen), Immunocytochemistry, Transcription Factor Activity and Detection
Clasificación Polyclonal
Concentración 0.25 mg/mL
Conjugado Unconjugated
Dilución Assay-dependent
Formulación PBS with 0.1% sodium azide; pH 7.4
génica PITX3
N.º de referencia del gen O35160
Alias de gen ak; aphakia; ASMD; ASOD; CTPP4; CTRCT11; Homeobox protein PITX3; homeobox protein PTX3; MGC12766; paired like homeodomain 3; paired-like homeodomain 3; paired-like homeodomain transcription factor 3; Pituitary homeobox 3; Pituitary homeobox 3 (Homeobox protein PITX3); PITX3; Ptx3
Símbolos de los genes PITX3
Especie del huésped Rabbit
Inmunógeno Synthetic peptide derived from the N-terminal region of the mouse PiTX3 protein.
Método de purificación Antigen affinity chromatography
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Neurobiology
Primario o secundario Primary
ID de gen (Entrez) 18742
Envase 1 Each
Incluye 0.1% Sodium azide
Especies diana Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.