Learn More
Piracetam-d8, CDN
Código de producto. 30323111
Cambiar vista
Click to view available options
Cantidad:
0.005 g
0.01 g
Tamaño de la unidad:
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
30323111 0.01 g 0.01 g
30325630 0.005 g 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30323111 Supplier CDN Supplier No. CDND73200.01G
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Descubra más de 4000 compuestos deuterados de alta calidad, con envío directo desde stock para mayor rapidez. Ofrecemos muchas API únicas, incluidos ingredientes farmacéuticos activos, nitrosoaminas, aminoácidos, esteroides, gases, pesticidas y ácidos grasos.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material Piracetam-d8
CAS 1329799-64-5
Fórmula molecular C6 D8 H2 N2 O2
Cantidad 0.01 g
Sinónimo 2,2-Dideuterio-2-(2,2,3,3,4,4-hexadeuterio-5-oxopyrrolidin-1-yl)acetamide, 2-Oxo-1-pyrrolidineacetamide-α,α,3,3,4,4,5,5-d8, 2-(2-Oxopyrrolidin-1-yl-3,3,4,4,5,5-d6)acetamide-2,2-d2, Piracetam-d8, Piracetam-D8
Fórmula InChI InChI=1 S/C6H10N2O2/c7-5(9)4-8-3-1-2-6(8)10/h1-4H2,(H2,7,9)/i1D2,2D2,3D2,4D2
SMILES [2 H]C([2 H])(N1C(=O)C([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H])C(=O)N
Nombre IUPAC 2,2-dideuterio-2-(2,2,3,3,4,4-hexadeuterio-5-oxopirrolidin-1-il)acetamida
Peso molecular (g/mol) 150.205
Formula Weight (peso de la fórmula) 150.124 g/mol
Porcentaje de pureza 98 atom % D, min 98% Chemical Purity