Learn More
Pigment Yellow 12 (Grado Técnico), TRC
Código de producto. 30012088
Cambiar vista
Click to view available options
Cantidad:
500 mg
1 g
5 g
Tamaño de la unidad:
1 g
5 g
500 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
30012088 1 g 1 g
30010579 5 g 5 g
30007056 500 mg 500 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 30012088 Proveedor Toronto Research Chemicals N.º de proveedor TRCP4378381G
Moléculas orgánicas de alta pureza y estándares analíticos, implementados estratégicamente en todo el mundo para impulsar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
CAS 6358-85-6
Fórmula molecular C32H26Cl2N6O4
Peso molecular (g/mol) 629.5
Sinónimo 2,2'-[(3,3'-Dichloro[1,1'-biphenyl]-4,4'-diyl)bis(2,1-diazenediyl)]bis[3-oxo-N-phenylbutanamide], 2,2'-[(3,3'-Dichloro[1,1'-biphenyl]-4,4'-diyl)bis(azo)]bis[3-oxo-N-phenylbutanamide, Amazon Yellow X 2485, Benzidine Lacquer Yellow G, Benzyl Yellow G, Bis(acetyl-N-phenylcarbamylmethyl)-4,4'-disazo-3,3'-dichlorobiphenyl, Brilliant Yellow Slurry, C.I. 21090, Carnelio Yellow GX, DY 728, Dainichi Benzidine Yellow G, Daltolite Fast Yellow GT, Diarylide Yellow, Diarylide Yellow AAA, Diarylide Yellow AAMX, Diarylide Yellow YT 553D, Disazo Yellow, EP 500, Eljon Yellow BG, Graphtol Yellow A-HG, Hancock Yellow 10010, Helio Yellow GW, Helio Yellow GWN, Irgalite Yellow BO, Irgalite Yellow BST, Irgalite Yellow BTR, Isol Benzidine Yellow G, Isol Benzidine Yellow G 2537, Isol Benzidine Yellow G Special, Japan Yellow 205, Japan Yellow No. 205, KET Yellow 406, Kromon Yellow GXT Conc, Kromon Yellow MTB, Laibo 178-1, Light Yellow JB;Light Yellow JBO, Light Yellow JBT, Lionol Yellow 1206, Lionol Yellow 1235P;Lionol Yellow 1243P, Lionol Yellow 1245P, Lionol Yellow 2RN, Lionol Yellow GGT;Lionol Yellow GR, Lionol Yellow SS-OW, Lionol Yellow TCH 1205, Lionol Yellow TT 1206, Lodestone Yellow YB 57, Lutetia Yellow 2JRST, Monolite Yellow 2GRA, NSC 521237, PY 12, Permanent Yellow DHG, Rangoon Yellow, Recolite Yellow BG, Recolite Yellow BGT, Sanyo Benzidine Yellow B, Sanyo Pigment Yellow GRT, Segnale Light Yellow 2GR, Segnale Light Yellow 2GRT, Segnale Yellow HG, Seikafast Yellow 2300, Seikafast Yellow 7055, Seikafast Yellow H 7055, Siloton Yellow 3GX, Siloton Yellow GTX;Sumikaprint Yellow 3A-O, Sun Yellow AAA, Sunbrite Yellow 273-3556, Sunbrite Yellow AAA, Symuler Fast Yellow 219, Symuler Fast Yellow 224, Verona Yellow X 1791, Vulcafor Fast Yellow GT, Vulcafor Fast Yellow GTA, Vulcol Fast Yellow GR, Yellow 205, Yellow No. 205;
Nombre IUPAC 2-[(E)-[4-[4-[(E)-(1-anilino-1,3-dioxobutan-2-il)diazenil]-3-clorofenilo]-2-clorofenil]diazenil-3-oxo-N-fenilbutanamida
SMILES CC(=O)C(N=Nc1ccc(cc1Cl)c2ccc(N=NC(C(=O)C)C(=O)Nc3ccccc3)c(Cl)c2)C(=O)Nc4ccccc4
Especificaciones
Nombre del producto químico o material Pigment Yellow 12 (Technical Grade)
Cantidad 1 g
Fórmula InChI InChI=1S/C32H26Cl2N6O4/c1-19(41)29(31(43)35-23-9-5-3-6-10-23)39-37-27-15-13-21(17-25(27)33)22-14-16-28(26(34)18-22)38-40-30(20(2)42)32(44)36-24-11-7-4-8-12-24/h3-18,29-30H,1-2H3,(H,35,43)(H,36,44)
Formula Weight (peso de la fórmula) 628.1393