Learn More
Picloram, 95%, Thermo Scientific™
Belongs to the puridine class of the auxinic compounds similar to the hormone Indole-3-acetic acid (IAA)
Marca: Thermo Scientific Alfa Aesar J66702.06
| Cantidad | 5g |
|---|
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Especificaciones
| Picloram | |
| 421°C | |
| Chlorine-like | |
| MFCD00012101 | |
| 479075 | |
| picloram, 4-amino-3,5,6-trichloropicolinic acid, tordon, borolin, grazon, amdon, 2-pyridinecarboxylic acid, 4-amino-3,5,6-trichloro, k-pin, tordon 10k, tordon 22k | |
| NQQVFXUMIDALNH-UHFFFAOYSA-N | |
| 4-amino-3,5,6-trichloropyridine-2-carboxylic acid | |
| 15965 | |
| 241.46 |
| 1918-02-1 | |
| >110°C (230°F) | |
| C6H3Cl3N2O2 | |
| 5g | |
| 14,7397 | |
| Soluble in acetone | |
| C1(=C(C(=NC(=C1Cl)Cl)C(=O)O)Cl)N | |
| 241.45 | |
| CHEBI:34922 | |
| 95% |