Learn More
Tocris Bioscience™ Phototrexate
Descripción
Phototrexate is a photoswitchable potent inhibitor of human dihydrofolate reductase (DHFR). Phototrexate is a photochromic analog of Methotrexate (Cat. No. 1230). Antifolate activity is switched on (cis-Phototrexate) by UVA (375 nm) light; it can be switched off (trans-Phototrexate) by blue or white light and by thermal relaxation in the dark. cis-Phototrexate (IC50 = 6 nM) significantly reduces the viability of HeLa cells compared with trans-Phototrexate (IC50 = 34 μM).
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-[4-[(1 E)-2-(2,4-Diamino-6-quinazolinyl)diazenyl]benzoyl]-L-glutamic acid |
| CAS | 2268033-83-4 |
| Cantidad | 5 mg |
| Objetivo | Dihydrofolate Reductase Inhibitors |
| Fórmula molecular | C20H19N7O5 |
| Pureza | 95% |