missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-ASAP2 (Tyr724) Polyclonal Antibody

Descripción
Phospho-ASAP2 (Tyr724) Polyclonal Antibody for Western Blot
ASAP2 is a multidomain protein belonging to the beta family with two ANK repeats, an Arf-GAP domain, a PH domain and a SH3 domain. This protein localizes in the Golgi apparatus and also at the plasma membrane where it colocalizes with protein tyrosine kinase 2-beta (PYK2) and forms a stable complex with PYK2 in vivo. ASAP2 is tyrosine phosphorylated by activated PYK2. It functions as a substrate and downstream target for PYK2 and SRC, a pathway that may be involved in the regulation of vesicular transport. ASAP2 activates the small GTPases ARF1, ARF5 and ARF6 and regulates the formation of post-Golgi vesicles and modulates constitutive secretion. It modulates phagocytosis mediated by Fc gamma receptor and ARF6. Usually detected in heart, brain, placenta, kidney, monocytes and pancreas, it is up-regulated during monocyte maturation. Multiple transcript variants encoding different isoforms have been found.
Especificaciones
Especificaciones
| Antígeno | Phospho-ASAP2 (Tyr724) |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Formulación | Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 |
| génica | ASAP2 |
| N.º de referencia del gen | O43150 |
| Alias de gen | 6530401G17Rik; AMAP2; Arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 2; ArfGAP with SH3 domain, ankyrin repeat and PH domain 2; Asap2; centaurin, beta 3; CENTB3; DDEF2; development and differentiation enhancing factor 2; development and differentiation-enhancing factor 2; Gm1523; Gm592; KIAA0400; PAG3; PAP; Pap-alpha; PAPbeta; Paxillin-associated protein with ARF GAP activity 3; PYK2 C terminus-associated protein; pyk2 C-terminus-associated protein; SHAG1 |
| Símbolos de los genes | ASAP2 |
| Especie del huésped | Rabbit |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?