missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-MSK1/MSK2 (Ser376/Ser360) Polyclonal Antibody
GREENER_CHOICE

Código de producto. 30536024
Change view
Click to view available options
Cantidad:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad
30536024 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30536024 Proveedor Invitrogen™ N.º de proveedor PA5148474

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

MSK1 is a mitogen and stress activated protein kinase 1 which belongs to the AGC family of kinases and is related in structure to the ribosomal p70 S6 kinase subfamily. MSK1 can be activated by ERK1/2 and SAPK2/p38 MAP kinase. It is also known to be required for the phosphorylation of CREB, ATF1 H3 and HMG14 in response to mitogen and stress. Similar to RSK, MSK1 contains two kinase domains (N term and a C term). Once phosphorylated on Thr581 and Ser360 by ERK1/2 and SAPK2/p38, MSK1 autophosphorylate on at least 5 sites. Of these autophosphorylation sites Ser212 and Ser376 get phosphorylated by the C terminal kinase domain of MSK1 which is essential for the catalytic activity of the N terminal kinase domain. MSK2 plays an essential role in the control of RELA transcriptional activity in response to TNF. Phosphorylates 'Ser-10' of histone H3 in response to mitogenics, stress stimuli and EGF, which results in the transcriptional activation of several immediate early genes, including proto-oncogenes c-fos/FOS and c-jun/JUN. May also phosphorylate 'Ser-28' of histone H3. Mediates the mitogen- and stress-induced phosphorylation of high mobility group protein 1 (HMGN1/HMG14). In lipopolysaccharide-stimulated primary macrophages, acts downstream of the Toll-like receptor TLR4 to limit the production of pro-inflammatory cytokines. Functions probably by inducing transcription of the MAP kinase phosphatase DUSP1 and the anti-inflammatory cytokine interleukin 10 (IL10), via CREB1 and ATF1 transcription factors.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Phospho-MSK1/MSK2 (Ser376/Ser360)
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.2 mg/mL
Conjugado Unconjugated
Formulación PBS with 5% trehalose and no preservative
génica Rps6ka5
N.º de referencia del gen O75582, O75676
Alias de gen 1110069D02Rik; 3110005L17Rik; 6330404E13Rik; 90 kDa ribosomal protein S6 kinase 4; 90 kDa ribosomal protein S6 kinase 5; 90kDa; AI848992; AI854034; Ccdc88b; coiled-coil domain containing 88B; mitogen and stress-activated protein kinase 1; mitogen- and stress-activated protein kinase 2; mitogen- and stress-activated protein kinase-2; mMSK2; MSK1; MSK2; MSPK1; nuclear mitogen- and stress-activated protein kinase 1; Nuclear mitogen- and stress-activated protein kinase 2; ribosomal protein kinase B; ribosomal protein S6 kinase A4; ribosomal protein S6 kinase A5; ribosomal protein S6 kinase alpha 4; ribosomal protein S6 kinase alpha-4; Ribosomal protein S6 kinase alpha-5; ribosomal protein S6 kinase, 90kD, polypeptide 4; ribosomal protein S6 kinase, 90kDa, polypeptide 4; ribosomal protein S6 kinase, 90kDa, polypeptide 5; ribosomal protein S6 kinase, polypeptide 4; ribosomal protein S6 kinase, polypeptide 5; RLPK; RLSK; RPS6KA4; RPS6KA5; RSKB; RSK-B; RSKL; RSK-like protein kinase; S6K-alpha-4; S6K-alpha-5
Símbolos de los genes Rps6ka4, Rps6ka5
Especie del huésped Rabbit
Inmunógeno Phosphopeptide containing human MSK1 S376 site.
Método de purificación Antigen affinity chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 8986, 9252
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.