missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Ephrin B1 (Tyr313, Tyr317) Polyclonal Antibody

Descripción
Antibody detects endogenous levels of EFNB1 only when phosphorylated at Tyr313/317.
Cell surface transmembrane ligand for Eph receptors, a family of receptor tyrosine kinases which are crucial for migration, repulsion and adhesion during neuronal, vascular and epithelial development. Binds promiscuously Eph receptors residing on adjacent cells, leading to contact-dependent bidirectional signaling into neighboring cells. The signaling pathway downstream of the receptor is referred to as forward signaling while the signaling pathway downstream of the ephrin ligand is referred to as reverse signaling. Binds to receptor tyrosine kinase including EPHA4, EPHA3 and EPHB4. Together with EPHB4 plays a central role in heart morphogenesis and angiogenesis through regulation of cell adhesion and cell migration. EPHB4-mediated forward signaling controls cellular repulsion and segregation from EFNB2-expressing cells. May play a role in constraining the orientation of longitudinally projecting axons.
Especificaciones
Especificaciones
| Antígeno | Phospho-Ephrin B1 (Tyr313, Tyr317) |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | EFNB1 |
| N.º de referencia del gen | P52795, P52796, P98172 |
| Alias de gen | CEK5 ligand; CEK5 receptor ligand; CEK5-L; CEL5-L; CFND; CFNS; EFB1; EFL3; EFL-3; EFL6; EFNB1; EFNB3; ELK ligand; ELK-L; EPH-related receptor tyrosine kinase ligand 2; ephrin B1; Ephrin B3; EphrinB1; ephrin-B1; Ephrin-B1 C-terminal fragment; Ephrin-B1 CTF; Ephrin-B1 ICD; Ephrin-B1 intracellular domain; Ephrin-B1 precursor; Epl2; EPLG2; EPLG8; kinase ephrin-B1; Lerk2; LERK-2; LERK8; Stimulated by retinoic acid gene 1 protein; Stra1; STRA1 protein; transmembrane ligand |
| Símbolos de los genes | EFNB1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?