Learn More
Invitrogen™ Phospho-eIF2b epsilon (Ser539) Polyclonal Antibody
Código de producto. 10586263
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
cada uno
10586263 100 μL cada uno
1 options
Código de producto. 10586263 Proveedor Invitrogen™ N.º de proveedor 44530G
Rabbit Polyclonal Antibody
Phospho-eIF2b epsilon (Ser539) Polyclonal Antibody for Western Blot
This gene encodes one of five subunits of eukaryotic translation initiation factor 2B, a GTP exchange factor for eukaryotic initiation factor 2 and an essential regulator for protein synthesis. Mutations in this gene and the genes encoding other EIF2B subunits have been associated with leukoencephalopathy with vanishing white matter.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Phospho-eIF2b epsilon (Ser539)
Aplicaciones Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Formulación Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3
génica Eif2b5
N.º de referencia del gen Q13144, Q64350, Q8CHW4
Alias de gen C81315; CACH; CLE; EIF-2B; eIF-2B GDP-GTP exchange factor subunit epsilon; EIF2B5; EIF2BE; EIF2Bepsilon; eukaryotic translation initiation factor 2B subunit epsilon; eukaryotic translation initiation factor 2B, subunit 5 epsilon; eukaryotic translation initiation factor 2B, subunit 5 epsilon, 82kDa; initiation factor eIF-2Be; LVWM; Translation initiation factor eIF-2B subunit epsilon
Símbolos de los genes Eif2b5
Especie del huésped Rabbit
Inmunógeno The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of rat eIF2Be that contains serine 535 (corresponding to serine 539 in the human sequence).
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 192234, 224045, 8893
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG